Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Corning® LSE™ Compact Centrifuge 120 V, Corning

Supplier: Corning
Corning®
6755
89204-722CS 3602.68 USD InStock
89204-722
Corning® LSE™ Compact Centrifuge 120 V, Corning
Centrifuges Benchtop Centrifuges
The Corning® LSE™ Compact Centrifuge is a space saving centrifuge ideal for use in Life Science and Industrial Research labs

The centrifuge's control panel features a large digital display and a convenient turn knob for setting parameter values. The unit’s microprocessor maintains precise control over operation with ten levels of acceleration and deceleration and a speed range from 200 rpm to 6,000 rpm. The LSE™ Compact Centrifuge has a maximum g-force of 4,185 x g. Speed can be set or displayed in rpm or g-force with both actual and pre-set values visible at all times.

The centrifuge is equipped with an electronic lid lock system, over speed protection, and an automatic shut off imbalance detection system. The Corning® LSE™ Compact centrifuge is cold room compatible.

Three angle rotors are available for use with the Corning® LSE™ Compact Centrifuge. The 15 mL rotor has a capacity for 12 tubes while the 50 mL rotor holds 6. Both rotors accept round and conical bottom tubes and include removable, autoclavable tube sleeves. The third angle rotor accepts 18 x 1.5/2.0 mL microcentrifuge tubes. In addition the LSE™ Compact Centrifuge will also accommodate a swing-out rotor for 6 x 5 mL. Optional adapters accommodate various tube sizes including standard conical tubes and round bottom blood tubes.

Certifications: CE Listed. CSA Compatible.

Ordering information: 230V models are also available.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty