Would you like to log in again?
Ion Source Cleaning Powder, Restek
Supplier: Restek
22685
89205-676EA 47.37 USD InStock
89205-676
Ion Source Cleaning Powder, Restek
Cleaning Agents Detergents and Surface Cleansers
Use this aluminum oxide powder to clean surfaces that contact the sample or ion beam when you encounter poor sensitivity and inadequate abundances at high masses.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty