Would you like to log in again?
OPTI-LYNX™ Trap Cartridges
Supplier: Optimize Technologies
OPTI-LYNX™
11-02874-TA 11-02872-DQ 11-03787-TK 11-02868-DQ 11-02870-TA 11-02868-TG 11-02872-TE 11-02868-TH 11-03787-DQ 11-02868-TK 11-02872-TG 11-03787-TL 11-02870-TH 11-02870-TK 11-02874-DQ 11-02874-TB 11-02874-TF 11-02874-TE 11-02870-DQ 11-02872-TK 11-02874-TK 11-02872-TL 11-02870-TB 11-02870-TE 11-02872-TB 11-02872-TD 11-02870-TD 11-02868-TA 11-03787-TG 11-02868-TB 11-02872-TH 11-02868-TL 11-02870-TF 11-02874-TD 11-03787-TA 11-03787-TD 11-02868-TE 11-03787-TF 11-02872-TA 11-03787-TE 11-02872-TF 11-02874-TH 11-03787-TB 11-02874-TL 11-02868-TD 11-02870-TL 11-02870-TG 11-02868-TF 11-03787-TH 11-02874-TG
89207-652PK 1270.38 USD InStock
89207-652 89207-650 89207-648 89522-288 83008-226 89207-646 89207-644 89522-284 83008-228 89522-286 89522-280 89522-282 89207-664 89207-662 83008-240 89207-660 89522-306 89522-308 83008-234 83008-232 89207-658 89522-276 89207-656 83008-238 89522-278 89207-654 89522-272 89522-274 89522-270 89207-674 89207-672 89207-670 83008-244 89522-302 89522-304 89207-668 89207-666 83008-246 89522-300 89207-686 89207-684 89207-682 89207-680 89207-678 89522-298 89207-676 89522-294 89522-296 89522-290 89522-292
OPTI-LYNX™ Trap Cartridges
Chromatography Columns
OPTI-LYNX™ trap cartridges combine the convenience of our quick-connect holder with a versatile selection of packed-bed cartridges to offer chromatographers numerous options for on-line or off-line sample clean-up and pre-concentration.
  • Rated to 6000 psi
  • Bed volumes from 4 to 100 µl (perfect for large volume trapping applications)
  • Can be loaded and regenerated
  • Cartridges share the same external dimensions
  • Can be used in on-line and off-line applications
Each cartridge shares the same external dimensions, allowing you to use different diameters and porosities with the same holder hardware. OPTI-LYNX™ traps are available in packed-bed volumes of 4 to 100 µl with a wide range of standard and custom packings. Whether you want to separate a peptide digest from its matrix for further analyses, or prepare a dilute small molecule sample for LC injection without any sample loss - OPTI-LYNX™ traps are the ideal tool to optimize your technique. These columns can be loaded and regenerated repeatedly for maximum value.
Ordering information: Custom packing available.
Delivery information: Includes five disposable trap cartridges.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty