Would you like to log in again?
EXP® Trap Cartridges Packed with 3 μm EXP Packings
Supplier: Optimize Technologies
EXP®
15-03969-HM 15-03964-HJ 15-03978-HK 15-03973-HG 15-03983-HH 15-03983-HK 15-03978-HG 15-03978-HH 15-03983-HI 15-03964-HL 15-03969-HI 15-03978-HM 15-03973-HM 15-03973-HJ 15-03978-HJ 15-03969-HG 15-03964-HG 15-03969-HL 15-03973-HH 15-03964-HI 15-03983-HG 15-03978-HL 15-03969-HH 15-03983-HJ 15-03969-HK 15-03964-HH 15-03973-HL 15-03969-HJ 15-03964-HM 15-03964-HK 15-03973-HI 15-03978-HI 15-03973-HK 15-03983-HL 15-03983-HM
89208-118PK 1068.41 USD InStock
89208-118 89208-136 89208-158 89208-116 89208-138 89208-180 89208-182 89208-160 89208-144 89208-122 89208-188 89208-166 89208-124 89208-168 89208-184 89208-140 89208-164 89208-142 89208-120 89208-186 89208-148 89208-126 89208-128 89208-190 89208-170 89208-192 89208-176 89208-154 89208-132 89208-134 89208-156 89208-172 89208-150 89208-174 89208-152
EXP® Trap Cartridges Packed with 3 μm EXP Packings
Chromatography Columns
The patented hand-tight EXP® trap column is rated for use at 20000+ psi (1400+ bar). This unique design connects directly to any injection valve (with 10-32 threads) or in-line with ¹/₁₆" stainless tubing for unparalleled convenience and efficiency.
  • Rated to 20000+ psi (1400+ bar)
  • Bed volumes from 1.6 to 600 μl
  • Convenient means of handling a variety of sample clean-up and enrichment processes
  • More efficient than off-line trapping techniques
  • Hand-tight trap replacement - no tools required
The EXP® cartridge system enables chemists to quickly remove detergents or salts which can affect the ionization process in MS work. This trapping technique can concentrate the sample directly on-line, allowing for increased recovery of precious sample material compared to off-line techniques. On-line trapping readily lends itself to automation for high-throughput analysis in UHPLC/MS applications. Free-Turn® architecture allows the user to change cartridges by hand without breaking fluid connections on the holder inlet/outlet.
Ordering information: Custom packing available.
Delivery information: Includes three uni-directional cartridges.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty