Would you like to log in again?
DeSCAL, Acidic Detergent, Decon Labs
Supplier: Decon Labs
DeSCAL
4001 4004 4004 4010 4001
89233-170CS 482.87 USD InStock
89233-170 CA89233-174 CA89233-172 89233-172 CA89233-170
DeSCAL, Acidic Detergent, Decon Labs
Cleaning Agents Detergents and Surface Cleansers
DeSCAL is a concentrated acidic detergent and descaling agent for removing hard water salt deposits from waterbaths, ultrasonic tanks, automatic labware washing machines, autoclaves, stills, labware, glass, plastics, and metals.
  • Dissolves calcium, magnesium, sodium, and iron deposits / just about every salt left behind from hard water
  • Powerful enough to remove crystallized urine deposits from animal cages
DeSCAL’s non-foaming formulation allows its use in automatic washers and spraying equipment.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty