Would you like to log in again?
Target DP™ Innovative Closure Technology, Thermo Scientific
Supplier: Thermo Scientific
C5000-53B C5000-44B C5000-50B C5000-54B C5000-55B C5000-99B C5000-51B C5000-50 C5000-45B
89239-426PK 81.37 USD InStock
89239-426 89239-424 89239-428 89239-418 89239-430 89239-422 89239-434 89239-420 89239-432
Target DP™ Innovative Closure Technology, Thermo Scientific
Closures Chromatography Vial Closures
The National Target™ DP™ 9mm C5000 series vial closure system is an innovative, flexible, and performance-driven system built on the Advance Vial Closure System (AVCS) technology platform.
  • No Septa Push-Through Due to AVCS Technology
  • Unlimited Options in Selecting the Best Septum for your Instrument and Applications
  • Optimized Ergonomics, Fine Texturing, and Evenly Spaced Ribbing for Easy Handling
These closures are designed to help chromatographers achieve more reliable and efficient sample analysis. Designed to accommodate nearly all chromatography autosamplers fitted for 12x32cm vial trays, the C5000 series closures effectively eliminate septa push-through and significantly enhance productivity and flexibility in the laboratory. The C5000 series closures are fully compatible with all of the C4000 series Target™ DP™ 9mm vials.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty