Would you like to log in again?
Phosphate Buffered Saline (PBS) 20X, Genetex
Supplier: GeneTex
GTX48888
89261-496EA 153.02 USD InStock
89261-496
Phosphate Buffered Saline (PBS) 20X, Genetex
Buffers Electrophoresis Buffers Washing Buffers
This solution is sufficient to prepare 10 L of 1X phosphate buffered saline
Tested applications: ELISA, FACS, ICC/IF, IHC, WB.
At 25 °C, 1X PBS will have a pH between 7.4 and 7.6. Not intended for any diagnostic or therapeutic purpose. Not suitable for human or animal consumption. Store at room temperature.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty