Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FTO Mouse Monoclonal Antibody [clone: 5-2H10]

Supplier: GeneTex
GTX82693
89284-130EA 675.09 USD InStock
89284-130
Anti-FTO Mouse Monoclonal Antibody [clone: 5-2H10]
Antibodies
Mouse monoclonal antibody [5-2H10] to Fatso

Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: FTO
Clonality: Monoclonal
Clone: 5-2H10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty