Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eppendorf® 5810 R Refrigerated Benchtop Centrifuge

Supplier: Eppendorf
022628180 022628092 022628258 022628187 022628257 022627082 022628089 022628089 022628168 022628169 022628180 022625101 022625101 022628181 022628169 022627133 022627122 022628187 022628188 022627104 022625501 022628257 022628258 022628092 022627122 022628067 022628188 022625551 022628181 022627040 022625501 022627023 022628067 022627133
89213-142EA 21844.32 USD InStock
CA11017-372 89213-142 89213-144 80076-424 CA89202-666 97007-374 89305-180 CA53513-812 89305-184 10856-382 CA11017-368 89210-990 89210-994 89210-992 CA11017-360 CA11017-362 CA89213-142 CA11017-366 CA80076-424 CA89213-144 97007-378 89202-666 89202-668 CA89202-668 CA89210-994 97007-382 53513-814 89210-988 97007-384 89305-172 53513-812 89305-174 CA89210-992 CA89210-990
Eppendorf® 5810 R Refrigerated Benchtop Centrifuge
Centrifuges Benchtop Centrifuges
The refrigerated Eppendorf Centrifuge 5810 R is the perfect day-to-day centrifuge for laboratories with medium-throughput requirements. It has an extremely compact footprint yet still offers the real versatility of being able to take both tube and plate formats.

  • Centrifuge lid with soft-touch lid closure
  • Low access height of 29 cm for easy loading and unloading of samples
  • Quiet operation to improve work environment
  • Compact footprint saves valuable bench space
  • Automatic rotor recognition and imbalance detection for maximum operational safety
  • Aerosol-tight Eppendorf QuickLock® caps and lids for easy, one-handed operation
  • Swing-bucket rotors and adapters accommodate tubes and bottles from 0.2 to 750 ml
  • Plate rotors for centrifugation of all types of MTP, PCR, cell culture, or Deepwell Plates
  • Fixed-angle rotors for high-speed applications in tubes from 0.2 to 85 ml
  • FastTemp function for fast pre-cooling
  • Continuous cooling maintains a constant temperature after the run has been completed - samples stay cool
  • ECO shut-off engages after 8 hours of non-use to reduce energy consumption and to extend compressor life
  • Dynamic compressor control (DCC) technology for optimized cooling performance

Refrigerated Centrifuge 5810 R, with its renowned quality and reliability, offers the most cost-effective solution for medium to high-throughput applications requiring speeds of up to 20,913 ×g (14,000 rpm). This benchtop centrifuge has been designed to fit into every laboratory and make routine work more comfortable thanks to its quiet operation, soft-touch lid closure and low access height for easy loading and unloading.

Experience true versatility with a broad selection of rotors and adapters that facilitate mixed loading of tubes, bottles and plates. A new generation of rotors increases the capacity of Centrifuge 5810 R to a maximum of 4×750 ml (or 28×50/56×15 ml). The swing-bucket rotor and adapters accommodate tubes and bottles from 0.2 to 750 ml, and the fixed-angle rotor 0.2 to 85 ml tubes. The plate rotor is ready to carry out centrifugation of all types of multiwell plates, such as PCR, cell culture, or deep-well plates.

Max. Capacity Plates: 16×MTP/4×DWP
Rotors Available: 18

Certifications: UL Listed, Certificate of Quality/Conformity (ISO 9001, ISO 13485, ISO 14001).

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty