Various adapters, rotors, and other accessories for centrifuges 5804 and/or 5804R and/or 5810 and/or 5810R. Eppendorf Rotors for Centrifuges 5804(R) and 5810(R) precision meets power. Customize your centrifugation with varied capacities and speeds. Accessorize with versatile buckets and adaptors for efficient sample processing. Ideal for high-performance applications, ensuring accuracy and reproducibility.
- Precision-engineered for optimal sample separation
- All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 min)
- Accessories (adaptors and buckets) further customize your centrifuge setup
- Aerosol-tight options available
Experience precision in high-performance centrifugation with Eppendorf Rotors designed for Centrifuge 5804/5804 R. These rotors offer diverse capacities and speeds, providing flexibility for various sample processing needs. Accessorize with versatile buckets and adaptors, enhancing the adaptability of your centrifuge setup. Ideal for high-performance applications, the Centrifuge 5804 series is further optimized with these precision-engineered rotors, ensuring accuracy and reproducibility in every spin. Explore the range to elevate your centrifugation experience.
Ordering information: Rotors, adapters, and accessories are only for use with centrifuge models specified. Unless otherwise specified, rotors may be used with all centrifuge models.