Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dilu-Lok™ Dilution Vial with Hinged Cap

Supplier: Hardy Diagnostics
DiluLok™
D089 D590 D085 D050 D590 D099 D699 D290 D599 D080 D080 D690 D090 D299 D290 D089 D090 D599 D085 D099 D299 D699 D690
89407-106CS 616.03 USD InStock
89407-106 10145-966 CA89407-116 CA89407-106 89407-108 89407-102 CA89407-100 CA89407-112 89407-114 CA89407-102 CA89407-114 89407-116 CA89407-108 CA89423-880 89407-098 CA89407-096 89407-110 89407-100 CA89407-098 CA89407-110 76169-782 89407-112 89407-096
Dilu-Lok™ Dilution Vial with Hinged Cap
Media for Microbiology Dehydrated Media for Microbiology
The Dilu-Lok™ eliminates the time and money spent on preparing your dilution vials! Our pre-filled dilution vial is designed especially for the food, dairy, pharmaceutical, and water industries. The vials are filled to 25, 90, or 99 ml with a variety of buffering and enrichment solutions. Each lot is meticulously tested for sterility and fill volume.

  • Heavy duty: Vials are made of sturdy polypropylene with a leak-resistant seal that holds up even during air transport
  • Leak resistant: Re-engineered for an improved seal, easy-to-remove, perforated, shrink wrap ensures product integrity
  • Optically clear: vials are made of clarified polypropylene for clear viewing

Large opening: A wide 45mm opening accommodates most sample sizes. Each vial is clearly marked with a 90 and 99 ml fill line.
One-handed: Flip-top lid allows for easy one-handed opening and closing to make pipetting easier. Saves unnecessary motion! No awkward screw caps or plastic strips to tear off.
Variety: Available with peptone water, buffered peptone water, Butterfield's buffer, deionized water, phosphate buffer with MgCl, and an empty sterile container.

Industry: Breweries, wineries and distilleries, canabis QA testing, cosmetics, environmental, food safety

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty