Would you like to log in again?
ART® Non-Filter Reload System, Thermo Scientific
Supplier: Thermo Scientific ART
ART®
3101-RT 3502-05-RT 3551-05-RT 3102-RT 3501-05-RT 3522-RT 3102-05-RT 3771-05-RT 3521-RT 3522-05-RT 3501-RT 3771-RT 3521-05-RT 3101-05-RT 3552-RT 3772-RT
89413-228CS 563.8 USD InStock
89413-228 89413-306 89413-308 89413-280 89413-282 89413-250 89413-196 89413-230 89413-286 89413-200 89413-310 89413-244 89413-202 89413-224 89413-226 89413-304
ART® Non-Filter Reload System, Thermo Scientific
Pipette Tips
ART® portfolio is now more complete, making it easier than ever to find a tip to fit virtually any pipette or application in the laboratory.
The space saving, stackable tip packaging allows for tips to pass through offset holes in lower insert. The easy to use, blister sleeve guide rails align tip inserts for smooth transfer into the rack. Supplied in sterile packaging, the outer sleeve protects tips from exterior contamination while refilling inserts. The environmentally-friendly tip packaging option reduces plastic waste. The rigid insert design provides reliable tip loading into multichannel devices. All tips are traceable, with lot number and description printed on every insert.
Renowned for reliable quality, the ART® series of pipette tips is now offering a complete selection of Barrier, Non-Filter, and Specialty tips. All formats deliver proven quality, performance, and reliability, backed by the highest Thermo Scientific standards and worldwide support.
Certifications: RNase-, DNase-, DNA-, ATP-, and pyrogen-free.
Ordering information: An empty rack is required for Reload Tower use.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty