Would you like to log in again?
Distilling Cow Receiver, Glassco
Supplier: Glassco
024.475.01
89426-360EA 42.09 USD InStock
89426-360
Distilling Cow Receiver, Glassco
Distillation Components Distillation Receivers
Suitable for microscale application
Receiver features one female joint and four drip-tip male joints.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty