Would you like to log in again?
Automation Tips for Eppendorf® epMotion™ Automated Pipetting System, Thermo Scientific
Supplier: Thermo Scientific
967-262 965-261 965-262 966-021 967-261 966-262 965-021 967-021 966-261
89428-728CS 905.28 USD InStock
89428-728 89428-734 89428-724 89428-736 89428-726 89428-730 89428-720 89428-732 89428-722
Automation Tips for Eppendorf® epMotion™ Automated Pipetting System, Thermo Scientific
Pipette Tips
Thermo Scientific provides a solution for your Eppendorf epMotion robotic pipette tips.
These automation pipette tips have been designed, validated, and tested on an actual epMotion workstation to ensure equal performance without the need to modify your existing protocols and programs. Our low percentage Coefficient of Variation (%CV) ensures product consistency and performance from every lot. Each lot must pass a 15-point QC inspection including performance testing on our epMotion workstation before tips are released. All tips are certified RNase, DNase, DNA, endotoxin, bioburden, and pyrogen free.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty