Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
CritiCore Molded Sole Cleanroom Boots with ESD Stripe
Supplier: CritiCore Protective Wear
CritiCore
89495-813 89495-827 89495-815 89495-829 89495-831 89495-823 89495-821 89495-819 89495-817 89495-835 89495-825 89495-833
89495-819PK 82.39 USD InStock
89495-819 89495-829 89495-817 89495-823 89495-833 89495-821 89495-831 89495-827 89495-815 89495-825 89495-813 89495-835
CritiCore Molded Sole Cleanroom Boots with ESD Stripe
Workwear
With a unique anti-slip tread pattern featuring 30% more traction and a high sidewall, these reusable molded-sole cleanroom boots provide superior performance in both wet and dry environments.
  • Gray molded sole
  • 45.7cm (18") fabric uppers with tunneled elastic at ankle and top
  • Three male snaps and one female snap at top for size adjustment
  • One additional reinforced snap on back of boot for coverall leg attachment
  • Reinforced fabric band is sewn around the perimeter of the sidewall for added durability
  • Buckle includes color-coded webbing for easy size identification
  • All seams are laser-cut or pre-serged
  • Thread is silicon-free
  • 99% polyester, 1% carbon material
They are gamma sterilization stable and made with a tightly woven calendered fabric that has excellent particle control properties. Applications include pharmaceutical, bio-tech, and medical device manufacturing.
They are gamma sterilization stable and made with a tightly woven calendered fabric that has excellent particle control properties. Applications include pharmaceutical, bio-tech, and medical device manufacturing.
Ordering information: Cleanroom boots are available in white or navy, with sizes ranging from small to 3X-large. All cleanroom boots ship in a pack of 2. Extended lead time may be required. Contact your Avantor sales representative for more information.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty