Micrewtube™ Microcentrifuge tubes are ideal for freezer storage, boiling application and centrifugation, and will fit most standard microcentrifuge rotors.
- Made of polypropylene
- Can be used at temperatures from –196 to 121 °C
Low adhesion Micrewtubes™ Tubes are also available. The specially formulated polypropylene used to manufacture these tubes provides a low adhesion surface to obtain maximum sample yield.
Non-skirted tubes withstand high speed centrifugation up to 20000 ×g. Skirted tubes can withstand up to 17000 ×g. Available plain, graduated, or brown for light sensitive material. Available with or without molded ridges.
Certifications: Certified RNase-, DNase-, DNA- and Pyrogen-free.
Ordering information: Caps sold separately; please see T340 Series for caps.