Would you like to log in again?
Screw Cap Microcentrifuge Tubes and Caps, Globe Scientific
Supplier: Globe Scientific
111702 111732 111700 111720 111722 111692 111710 111742 111712 111730 111690 111740
89501-416PK 124.07 USD InStock
89501-416 89501-426 89501-418 89501-428 89501-412 89501-422 89501-414 89501-424 89501-430 89501-432 89501-410 89501-420
Screw Cap Microcentrifuge Tubes and Caps, Globe Scientific
Tubes Microcentrifuge Tubes
This line of disposable screwcap microcentrifuge tubes are available in sterile or non sterile formats.
  • Tubes and caps are manufactured from polypropylene
  • Maximum RCF for all tubes is 18000×g
Sterile tubes include a cap with nitrile O-Ring. Non sterile tubes and caps are sold separately, allowing a choice of cap styles (with or without O-ring), as well as color.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty