Would you like to log in again?
HYB-LINK™ Formamide-Free Hybridization Buffer, G-Biosciences
Supplier: G-Biosciences
786-405 786-406 786-405 786-406
95029-218EA 225.77 USD InStock
CA95029-220 95029-218 CA95029-218 95029-220
HYB-LINK™ Formamide-Free Hybridization Buffer, G-Biosciences
Buffers Electrophoresis Buffers Hybridization Buffers
G-Biosciences' Hyb-LINK™ is a unique, high performance, formamide-free hybridization buffer solution
G-Biosciences' Hyb-LINK™ provides high resolution and increases the sensitivity of random-primer DNA probes. Hyb-LINK™ capitalizes on increased sensitivity of blot hybridization without increasing the background, and therefore, provides reliable and accurate hybridization.
G-Biosciences' Hyb-LINK™ is suitable for all types of probes (compatible with both RNA and DNA probes). The protocol involves incubation of blots with denatured probes in Hyb-LINK™ followed by washing of the blot to remove excess unhybridized probes.
It is suitable for Southern, Northern, Southwestern and Dot Blot hybridization analysis, hybridization with oligonucleotides or other types of probes, and for hybridization screening of gene libraries. It provides researchers with unprecedented simplicity, reliability, accuracy for hybridization experiments.
Ordering information: Accessories available.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty