Would you like to log in again?
Electrophoresis Running Buffers, G-Biosciences
Supplier: G-Biosciences
786-474 786-482 786-480 R025 786-481 786-531 786-029 786-485 786-486 786-060 786-025 786-025 786-420 786-483 786-029G 786-029G 786-475 786-532 786-486 786-421 786-060 786-475 786-420 786-484 786-531 786-029 786-481 786-479 R025 786-482 786-483 786-532 786-480 786-484 786-474 786-485 786-701 786-421 786-472 786-479 786-472
82021-496EA 65.77 USD InStock
82021-496 CA82021-242 CA95043-526 10153-804 95029-270 95029-272 95029-250 82021-250 95029-252 95029-274 82021-252 95029-254 95029-276 CA95029-262 CA95029-276 CA95029-254 95029-266 95029-268 95029-224 95029-226 89167-720 95043-526 95043-524 82021-242 CA82021-252 CA82021-496 CA95029-270 CA95029-272 CA95029-250 95029-262 CA95029-252 CA95029-274 95029-264 CA82021-250 CA95029-264 CA95029-266 CA89167-720 CA95029-224 CA95029-268 CA95029-226 CA95043-524
Electrophoresis Running Buffers, G-Biosciences
Buffers Electrophoresis Buffers Running Buffers
G-Biosciences offers many electrophoresis running buffers including: TAE Running Buffer [50X], TBE Running Buffer [10X], TBE-Urea Sample Buffer, MOPS Running Buffer [10X], and MOPS/ SDS Running Buffer [20X]
  • The TAE Running Buffer [50X] contains 2M Tris-Acetate and 0.05M EDTA (at a pH of 8.3)
  • The TBE Running Buffer [10X] contains 0.9M Tris-borate and 0.01M EDTA (at a pH of 8.3)
  • The MOPS Running Buffer [20X] contains 200 mM MOPS (at a pH of 7.0), 80 mM Sodium acetate and 10 mM EDTA
G-Biosciences' TBE-Urea Sample Buffer is a loading buffer used to resolve single-stranded DNA oligos or RNA into sharp, distinct bands. The TBE-Urea Sample Buffer is for the analysis and purification of products ranging from 20-800 bases, making them an ideal choice for synthetic oligo analysis and purification, RNase Protection Assays (RPA), in vitro transcription studies, and Northern blot analysis.
These include the SDS-PAGE Sample Loading Buffer, SDS-PAGE Running Buffer, Native Sample Loading Buffer, Tris Glycine Native Gel Running Buffer, Tricine Sample Buffer, Tris-Tricine, Tris-Tricine-SDS, MES SDS Running Buffer, MOPS/ SDS Running Buffer, IEF Anode Buffer, IEF Cathode Buffer, Zymogram Sample Buffer, Zymogram Renature Buffer, Zymogram Development Buffer and the Discontinuous Buffer System.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty