Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSF1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
SPA-901F ADI-SPA-901-D ADI-SPA-901-J ADI-SPA-901-F SPA-901J SPA-901D
101410-578EA 233 USD InStock
101410-578 95045-164 CA101410-580 101410-580 CA95045-164 CA101410-578
Anti-HSF1 Rabbit Polyclonal Antibody
Antibodies
HSFs (Heat Shock family of transcription factors), which consists of HSF 1-4, bind to highly conserved Heat shock elements (HSEs) in the promoter regions of heat shock genes, ultimately regulating the expression of Heat shock proteins (Hsps). On exposure to heat shock and other stresses, HSF1 localizes within seconds to discrete nuclear granules and on recovery from stress, HSF1 rapidly dissipates from the stress granules to a diffuse nucleoplasmic distribution.

  • For ELISA, Flow Cytometry, Immunoprecipitation, Western Blot, EMSA

Recognizes human, mouse, rat, gerbil, monkey and rabbit HSF1. Detects a band of ~95kDa by Western blot.

Type: Primary
Antigen: HSF1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty