Would you like to log in again?
WHEATON® MicroLiter µLPlate® 96-Well Microplates, DWK Life Sciences
Supplier: DWK Life Sciences
WHEATON® µLplates®
07-2000-200 07-8000-1200 07-0000-C 07-1100 07-0063N 07-3000-350 07-8000 07-1000 07-0010N-B 07-3000 07-0011N-B 07-6000 07-8000-2200 07-0015N 07-2000-250 07-0010N 07-5000 07-0060N 07-7000 07-8000-1200-S 07-0066N 07-3000-300 07-0011N 07-2000-201 07-1200-101 07-7400-SI 07-7400 07-1200 07-8000-1202 07-0065N 07-3900 07-2400 07-0061Y 07-2100-B 07-80001000S 07-8000-1000 07-2100 07-0061N 07-0017N 07-0067N 07-8400
97044-562EA 13.58 USD InStock
97044-562 10806-064 97044-548 97044-944 97044-824 97045-190 97044-568 97044-542 97045-372 97045-374 97045-352 97045-870 97044-916 97045-172 97045-173 97045-332 97045-354 97045-376 97045-058 97045-378 97044-552 97044-550 97044-834 97044-832 97044-830 97044-930 89176-336 97044-554 97045-186 97045-364 97045-188 97045-342 97045-060 97044-828 97044-946 97044-826 30618-326 30618-204 97045-322 97045-366 97045-368
WHEATON® MicroLiter µLPlate® 96-Well Microplates, DWK Life Sciences
Microplates Deep Well Microplates
A comprehensive line of 96-well microplates for chromatography, the Plate Sampling System™ uses closures familiar to those who work with standard chromatography vials
Polypropylene μLplates™ have a variety of closures that will stabilize the sample during the analytical sample run and, if necessary, reseal after the sample has been injected. You may also choose the security of a stopper coupled with the strength of a mechanical closure system. The mechanical closure system permits even GC samples can be converted to 96-well format, by sealing the limited volume insert on the inside surface of the insert and compressing the PTFE/silicone rubber using a mechanical seal. A variety of insert options let you choose the specifications appropriate for your application.
The system combines microplates, glass inserts, μLmats™, µLmat Vise™ and caps with chromatography-based septa to offer you secure vessels to maintain even the most volatile samples
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty