Would you like to log in again?
CAPS SNAP TOP BLUE STAR PRESLT
Supplier: MicroSolv Technology Corp
Autosampler Blue snap top Cap^ With star preslit, ultra low bleed , white silicone/ yellow PTFE septa, 0.40inch thick. For use with 12x32mm snap top vials. Basik brand. 100/pack.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CAPS SNAP TOP BLUE STAR PRESLT 97055-296 9502C-E-31CB Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty