Would you like to log in again?
EXP® Trap Cartridges, 2.7 µm EXP
Supplier: Optimize Technologies
EXP®
15-03964-HA 15-03973-HD 15-03973-HN 15-03978-HF 15-03978-HB 15-03978-HN 15-03983-HA 15-03969-HD 15-03973-HB 15-03969-HA 15-03983-HD 15-03978-HA 15-03983-HE 15-03969-HB 15-03969-HE 15-03978-HD 15-03969-HN 15-03964-HB 15-03969-HF 15-03983-HN 15-03964-HF 15-03964-HD 15-03978-HE 15-03973-HE 15-03983-HF 15-03973-HA 15-03983-HB 15-03964-HN 15-03973-HF 15-03964-HE
89208-114PK 1182.94 USD InStock
89208-114 97056-318 97056-342 97056-320 97056-340 97056-306 97056-328 89208-146 97056-326 97056-304 89208-162 97056-302 97056-324 97056-300 97056-308 97056-298 97056-330 97056-296 97056-294 97056-292 97056-316 97056-338 97056-314 89208-178 97056-336 97056-334 97056-312 97056-332 89208-130 97056-310
EXP® Trap Cartridges, 2.7 µm EXP
Chromatography Columns
The patented hand-tight EXP® Trap Column is rated for use at 20,000+ psi (1,400+ bar). This unique design connects directly to any injection valve (with 10-32 threads) or in-line with ¹/₁₆" stainless tubing for unparalleled convenience and efficiency.
  • Rated to 20000+ psi (1400+ bar)
  • Bed volumes from 1.6 μl to 600 μl
  • Convenient means of handling a variety of sample clean-up and enrichment processes
  • More efficient than off-line trapping techniques
  • Hand-tight trap replacement - no tools required
The EXP® Cartridge System enables chemists to quickly remove detergents or salts which can affect the ionization process in MS work. This trapping technique can concentrate the sample directly on-line, allowing for increased recovery of precious sample material compared to off-line techniques. On-line trapping readily lends itself to automation for high-throughput analysis in UHPLC/MS applications. Free-Turn® architecture allows the user to change cartridges by hand without breaking fluid connections on the holder inlet/outlet.
Our specialized features, patented technology, precision engineering, and state-of-the-art manufacturing make the new EXP Trap Column and EXP Stem Trap unbeatable choices for UHPLC trapping applications.
Ordering information: Custom packing available.
Delivery information: Includes 3 uni-directional cartridges.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty