Would you like to log in again?
CAP FOR 40ML EPA VIALS SOLID
Supplier: MicroSolv Technology Corp
Caps, Screw^ Polypropylene white solid, closed top with a red butyl / grey PTFE septa. 55 shore A 2.5mm thick. Use with storage vials
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CAP FOR 40ML EPA VIALS SOLID 97057-240 97040-SP-24 Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty