Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diammonium hydrogen orthophosphate ≥98% ACS

Supplier: VWR
Synonyms: Diammonium hydrogen phosphateAmmonium PhosphateFyrexdi-Ammonium hydrogen phosphateDiammonium hydrogen orthophosphateAmmonium biphosphateAmmonium hydrogen phosphateAmmonium monohydrogen phosphateAmmonium phosphate dibasic

0569-500G 0569-1KG 0569-2.5KG
97061-930CS 908.88 USD InStock
97061-930 97061-928 97061-932
Diammonium hydrogen orthophosphate ≥98% ACS
di-Ammonium hydrogen orthophosphate


Formula: (NH₄)₂HPO₄
MW: 132.06 g/mol
Melting Pt: 185 °C
Density: 1.619 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010891
CAS Number: 7783-28-0
EINECS: 231-987-8
REACH: 01-2119490974-22
Merck Index: 12,00576
Order Now
Specification Test Results
Calcium 0.001%
Chloride = 0.001%
Heavy Metals (as Pb) = 0.001%
Insolubles = 0.005%
Iron = 0.001%
Magnesium = 0.0005%
Nitrate = 0.003%
pH (5%, Water) @25°C 7.7 - 8.1
Potassium = 0.005%
Purity = 98%
Sodium = 0.005%
Sulfate = 0.01%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty