Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tween® 20 (Polysorbate), Proteomics Grade

Supplier: VWR
Synonyms: Polyoxyethylene sorbitan monolaurate

M147-1L M147-4L
97063-874EA 128.45 USD InStock
97063-874 97063-872
Tween® 20 (Polysorbate), Proteomics Grade
Tween® 20 (Polysorbate)
Non-ionic detergent efficiently separates hydrophilic proteins from membrane spanning, hydrophobic proteins without altering biological activity.

  • High purity and protease-free
  • Suitable for most biological applications

Non-ionic detergents have uncharged hydrophilic groups and are gentle, non-denaturing detergents ideal for studying the conformation and function of proteins, because they keep secondary and tertiary structure intact.


Formula: C₅₈H₁₁₄O₂₆
MW: 1227.53 g/mol
Boiling Pt: > 100 °C
MDL Number: MFCD00165986
CAS Number: 9005-64-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty