Would you like to log in again?
Pfu DNA Polymerase, Agilent Technologies
Supplier: Agilent
600153 200532 600159 600140 200532 600154 600153 600160 600159 600160 600140 600136 600136 600154 600135 600135
99900-614EA 134.36 USD InStock
99900-614 99902-300 CA99902-298 97066-436 CA99900-614 CA99902-300 CA97066-436 99902-290 99900-166 CA99902-290 CA99900-164 99900-220 CA99900-166 CA99900-220 99902-298 99900-164
Pfu DNA Polymerase, Agilent Technologies
Nucleic Acid Reagents End-point PCR Enzymes and Kits
High-quality polymerases for difficult to amplify DNA targets.
  • Proofreading DNA polymerase isolated from Pyrococcus furiosus
  • Ideal choice for many techniques requiring high-fidelity DNA synthesis by PCR
  • Blunt ends
Pfu DNA Polymerases includes Pfu DNA Polymerase, a proofreading DNA polymerase isolated from Pyrococcus furiosus.
The portfolio also features the recombinant Exo- Pfu DNA Polymerase (a genetically engineered mutant of the cloned Pfu DNA format) and the PfuTurbo DNA Polymerase with an optional alternative detergent.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty