Would you like to log in again?
PfuTurbo DNA Polymerase, Agilent Technologies
Supplier: Agilent
600412 600410 600257 600256 600414 600256 600257 600602 600250 600259 600602 600254 600600 600252 600320 600252 600320 600324 600322 600412 600255 600414 600600 600322 600255 600324 600250 600259 600254 600410
97066-444EA 1133.13 USD InStock
CA99902-324 97066-444 CA97066-450 ST600600 CAST600250 ST600255 ST600257 ST600410 ST600250 ST600252 CAST600412 99902-380 CA99901-814 CAST600259 CAST600410 CAST600257 CAST600255 CAST600252 99902-324 CA97066-444 97066-450 ST600259 ST600412 ST600320 ST600322 CAST600600 CA99902-380 CAST600322 99901-814 CAST600320
PfuTurbo DNA Polymerase, Agilent Technologies
Nucleic Acid Reagents End-point PCR Enzymes and Kits
This DNA polymerase is ideal for a variety of techniques requiring high-fidelity DNA synthesis by the polymerase chain reaction (PCR).
PfuTurbo DNA Polymerase is an enhanced version of Pfu DNA Polymerase for robust, high-fidelity PCR. PfuTurbo DNA polymerase is a mixture of cloned Pfu DNA polymerase and the exclusive thermostable ArchaeMaxx polymerase-enhancing factor that enhances PCR product yields and increases target length capability without altering DNA replication fidelity.
PfuTurbo DNA Polymerase AD features a reformulated buffer system for increased economy with the same high performance as the original PfuTurbo DNA Polymerase for robust, high-fidelity PCR.
PfuTurbo Hotstart DNA Polymerase is a special formulation of high fidelity PfuTurbo DNA Polymerase with hot start activity.
PfuTurbo Cx Hotstart DNA Polymerase is a specially formulated with a mutant of Pfu DNA Polymerase designed to overcome uracil sensitivity for more robust performance.
The PfuTurbo Hotstart PCR Master Mix is a 2X formulation of PfuTurbo hotstart DNA Polymerase, an optimized PCR reaction buffer, magnesium, and dNTPs.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty