Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium dihydrogen phosphate monohydrate ≥99.998% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Sodium dihydrogen orthophosphate 1-hydrateSodium dihydrogen orthophosphate monohydrateSodium dihydrogen phosphate 1-hydrateSodium dihydrogenorthophosphate monohydrateSodium orthophosphate monobasic monohydrateSodium phosphate monobasic monohydrate

10870-14 10870-22 010870-14 010870-22
AA10870-14EA 145.99 USD InStock
AA10870-14 CAAA10870-14 AA10870-22 CAAA10870-22
Sodium dihydrogen phosphate monohydrate ≥99.998% (metals basis), Puratronic®
Sodium dihydrogen phosphate monohydrate


Formula: NaH₂PO₄·H₂O
MW: 137.99 g/mol
Melting Pt: 100 °C (-H₂O)
Density: 2.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149208
CAS Number: 10049-21-5
EINECS: 231-449-2
Merck Index: 13,08734
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/57/16640557.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty