These brass frame, stainless steel mesh sieves meet ASTM E11 requirements and all current amendments.
- Multiple diameters and mesh sizes available
- Mesh size is checked using a Nikon optical comparitor to confirm conformity with ASTM E11
- Superior quality sieves guarantee accuracy and reproducibility
- Sturdy construction enables easy stacking
All sieves are full height. The 7.62cm (3") diameter sieves are 2.54cm (1") deep. The 20.3cm (8") diameter sieves are 5.1cm (2") deep. The 30.5cm (12") diameter sieves are 7.62cm (3") deep. Sieve covers, made of polished brass, have a recessed wire handle are sold separately.