Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Citrate, 0.5 M Buffer Solutions, Thermo Scientific

Supplier: Thermo Scientific
J61690-AP J60125-AP J61249-AP J61249-AP J60754-AK J61391-AP J61690-AK J61690-AK J60024-AK J60754-AP J63008-AK J60125-AP J61391-AK J60024-AP J60754-AP J61249-AK J61690-AP J60125-AK J63008-AP J61391-AK J60754-AK J61249-AK J63008-AP J60125-AK J60024-AP J61391-AP J63008-AK J60024-AK
AAJ61391-AKEA 182.55 USD InStock
AAJ61391-AK CAAAJ63008-AK CAAAJ61391-AK CAAAJ63008-AP AAJ63008-AK AAJ61391-AP AAJ63008-AP CAAAJ61391-AP AAJ61690-AK CAAAJ60024-AP CAAAJ60754-AP AAJ60754-AK AAJ60024-AK CAAAJ61690-AP CAAAJ61690-AK AAJ60024-AP AAJ60125-AK CAAAJ60125-AP CAAAJ60754-AK AAJ60125-AP AAJ60754-AP CAAAJ60125-AK CAAAJ61249-AP AAJ61249-AP CAAAJ61249-AK AAJ61690-AP CAAAJ60024-AK AAJ61249-AK
Citrate, 0.5 M Buffer Solutions, Thermo Scientific
Buffers Biological Buffers
Colourless odourless buffer solutions.

  • CAS No.: 6132-04-3
  • MDL No.: MFCD00150031

Citrate, is used as an anticoagulant and also used as a biological buffer. It is often used to prepare sodium citrate buffer for antigen retrieval of tissue samples. The citrate solution is designed to break protein cross-links, thus, unmasking antigens and epitopes in formalin-fixed and paraffin embedded tissue sections, resulting in enhancing staining intensity of antibodies. Citrate has anticoagulant activity and as a calcium chelator, it forms complexes that disrupt the tendency of blood to clot.

Caution: Store in refrigerator. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty