Would you like to log in again?
Bond Elut C18, Agilent Technologies
Supplier: Agilent
Bond Elut™
52113024 14256001 14102052 14113027 52113024 52102052 12256023 12256031 12256001B 12256023 14102028 14113001 52102052 14102025 14102028B 14113027 14102028B 14113024 14256031 12105102B 52102024 14256031 14102001 14256015 12105027 14256001 12105027 14102025 14102028 14113024 14113001 14256015 14102001 12256031 14102052 52102001 12256001B 52102001 52102024
AG14102028BPK 1668.18 USD InStock
CAAG14256001 AG14102028B AG12105102B CAAG52102052 CAAG14102028B CAAG14102052 AG12105027 AG12256023 CAAG14113001 AG14256001 CAAG14113024 AG14102052 CAAG12256031 AG52113024 CAAG14113027 CAAG12256001B CAAG52102024 CAAG14102025 CAAG52102001 AG14113001 CAAG14102001 CAAG14256031 AG12256001B AG12256031 CA52113024 CAAG14256015 AG14113027 AG52102024 CAAG14102028 AG14113024 AG52102001 AG14256031 AG14102001 AG52102052 CAAG12256023 CAAG12105027 AG14102028 AG14102025 AG14256015
Bond Elut C18, Agilent Technologies
Chromatography Columns
Bond Elut C18 is a hydrophobic, end capped silica sorbent commonly used to extract nonpolar analytes.
  • The most hydrophobic, bonded silica sorbent
  • Extremely retentive for nonpolar compounds
  • Effective for desalting aqueous mixtures
Bond Elut C18 is the most hydrophobic, bonded silica sorbent available in the Bond Elut family. It is considered to have the broadest retention spectrum among bonded silica sorbents. C18 is the most popular SPE sorbent because of its extreme retention of nonpolar compounds.
Bond Elut C18 retains most organic analytes from aqueous matrices. When analyzing small to intermediate molecules, Bond Elut C18 can be used for desalting aqueous matrices before ion exchange, as salts pass through the sorbent unretained.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty