Would you like to log in again?
InfinityLab Well Plates, Agilent Technologies
Supplier: Agilent
InfinityLab
5065-4402 5043-9310 5043-9310 5043-9314 5043-9312 5043-9313 5043-9302 5043-9302 5043-9300 5043-9305 5043-9309 5043-9305 5043-9311 5043-9312 5043-9309 5043-9311 5043-9308 5043-9315 5065-4402 5043-9314 5043-9300 5043-9313 5043-9308 5043-9315
AG5043-9309PK 431.56 USD InStock
CA5065-4402 AG5043-9309 AG5065-4402 CAAG5043-9308 CAAG5043-9309 CA5043-9311 CA5043-9300 CA5043-9310 CA5043-9302 CAAG5043-9313 CA5043-9312 AG5043-9300 AG5043-9311 CAAG5043-9314 AG5043-9310 CAAG5043-9315 CA5043-9305 AG5043-9315 AG5043-9314 AG5043-9302 AG5043-9313 AG5043-9312 AG5043-9308 AG5043-9305
InfinityLab Well Plates, Agilent Technologies
Microplates Deep Well Microplates
A comprehensive selection of InfinityLab well plates and sealing mats for the HPLC workflow.
  • Ensure a perfect fit between well plates and slitted sealing mats
  • Access the right formats to ensure each well plate sampler setup is perfectly equipped for all HPLC applications
  • Optimized for high performance in your chromatography workflow
  • A large range of well volumes and package quantities provide the flexibility needed to suit your lab size and your needs
  • Compliant with worldwide ANSI and SLAS standards
Agilent provides a comprehensive selection of InfinityLab well plates and sealing mats for the HPLC workflow. 96 and 384 plates are available in different well sizes, well shapes, and bottom shapes to best suit your needs for all possible applications.
Choosing the right well plate can help you avoid such pitfalls as missed data points, sample rerun, and a failure to meet project deadlines. Agilent InfinityLab well plates and sealing mats for the HPLC workflow will help to produce the highest quality data for most common screening challenges.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty