Would you like to log in again?
CrossLab Cylindrical Cells for UV-Vis and UV-Vis-NIR Systems, Agilent Technologies
Supplier: Agilent
6618000600 5063-6566 6610002200 5063-6566 5061-3392 6610002300 5061-3392 6618000600 6610002200 6610002300
AG5063-6566EA 345 USD InStock
AG5063-6566 AG6618000600 CAAG5063-6566 CAAG5061-3392 AG5061-3392 AG6610002300 AG6610002200 CAAG6610002300 CA6618000600 CAAG6610002200
CrossLab Cylindrical Cells for UV-Vis and UV-Vis-NIR Systems, Agilent Technologies
Cuvettes Spectrophotometer Cuvettes
Agilent cylindrical cuvettes with 10 to 100 mm path length for both 8453/8454 and Cary UV systems.
  • Long pathlength, 2-port cylindrical cells provide extra sensitivity for low concentration samples
  • Micro cylindrical cells are for use with concentrated samples and help overcome high solvent absorbance (e.g. water in the NIR)
  • Requires a cylindrical cell holder for mounting in the optical path
  • Compatible with thermostatted cylindrical cell holder for added flexibility
  • All cylindrical cells are optimized for use with Agilent's market-leading portfolio of UV-Vis systems
Agilent's cylindrical cuvettes (cells) enhance performance and provide added flexibility for a range of applications with a variety of path length options available. All cyclindrical cells require use of a cylindrical cell holder and are compatible with thermostatting options for flexible and accurate performance.
For low concentration samples, the 2-port, long pathlength cylindrical cells offer extra sensitivity when required for clearer spectrophotometric results every time. For concentrated samples, or to help overcome high solvent absorbance, the short pathlength cylindrical cells are recommended.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty