Would you like to log in again?
CrossLab Column Selection Valves and Supplies for HPLC, Agilent Technologies
Supplier: Agilent
CrossLab
5068-0077 0101-1421 5067-4284 5068-0093 5068-0067 5067-4240 5067-4241 5068-0076 5067-4111 5068-0200 5067-4134 5067-4273 5068-0044 5067-4144 5068-0002 0101-1415 5067-4283 5067-4144 5068-0097 5068-0011 5067-4284 5068-0008 5067-4283 5068-0008 5068-0097 5068-0200 5067-4132 5067-4193 0101-1415 5068-0012 5067-4111 5067-4282 5068-0199 5068-0286 5068-0076 5067-6682 5068-0093 5067-4240 5067-4159 5068-0041 5067-4233 5067-4273 5068-0011 0101-1421 5068-0077 5067-4279 5067-6682 5068-0045 5067-4134 5067-4174 5068-0041 5067-4159 5068-0012 5068-0067 5068-0199 5068-0286 5068-0045 5068-0002 5068-0298 5067-4282 5067-4132 5067-4174 5067-4233 5067-4279 5067-4241 5068-0298 5068-0044 5067-4193
AG5068-0067EA 624.44 USD InStock
CAAG5068-0067 AG5068-0067 CA5068-0008 CAAG5067-4174 AG5067-4174 AG5067-4132 AG5067-4134 CAAG5067-4134 CAAG5067-4132 CAAG5068-0097 CAAG5068-0298 CAAG5068-0011 CA5068-0012 AG5068-0199 AG5068-0077 AG5068-0076 CAAG5068-0093 CAAG5067-4283 CAAG5067-4284 CAAG5067-4240 CAAG5067-4282 AG5067-6682 AG5067-4144 CAAG5067-4241 CAAG5068-0041 CAAG5068-0045 AG5068-0041 CAAG5068-0286 CAAG5068-0044 CAAG5068-0200 CAAG5068-0002 AG5068-0045 AG5068-0044 AG5068-0286 AG5068-0200 AG5068-0002 AG0101-1421 CAAG5067-4273 AG5068-0008 AG5067-4193 CAAG5067-4193 AG5067-4273 AG5067-4111 CAAG5067-4159 AG5067-4233 CAAG5067-4279 AG5067-4279 CAAG5067-4111 CA0101-1415 AG5067-4159 CAAG5067-4233 CAAG5068-0076 CAAG5068-0199 CAAG5068-0077 AG5068-0093 CA0101-1421 AG5068-0012 AG5068-0298 AG5068-0011 AG5068-0097 CAAG5067-6682 AG5067-4282 AG5067-4283 AG5067-4284 AG5067-4240 AG5067-4241 AG0101-1415 CAAG5067-4144
CrossLab Column Selection Valves and Supplies for HPLC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
High-pressure valve heads, rotor seals and stators, and bearing rings for column switching valves.
  • InfinityLab Quick Change Valves specially designed specifically for Agilent HPLC systems
  • Greater flexibility in solvent and column selection, new automation capabilities, and increased sample throughput as well as improved separation performance
  • Rotor seals made from different materials such as PEEK, Vespel, or Tefzel for different applications and pH ranges
Agilent provides a set of column switching valves, as well as kits and supplies, for enhanced TCC performance and applications. Available supplies include high-pressure valve heads with integrated RFID tags, replacement rotor seals and stators, and bearing rings/isolation rings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty