Would you like to log in again?
CrossLab Manual Injection Valves and Supplies for HPLC, Agilent Technologies
Supplier: Agilent
CrossLab
5067-7031 1535-4048 1400-3166 5068-0321 0101-1243 5067-1503 0101-0376 0101-1248 5067-1503 5067-6831 5067-7032 1400-3166 0101-1243 1535-4048 0101-1242 1535-4046 5067-7033 0101-1248 5067-7031 5063-6502 1535-4046 5067-4191 0101-1245 5067-7033 5067-4191 0101-1244 0101-1242 5067-7032 0101-0376 5065-9922 0101-1246 5067-6845 0101-1245 5068-0321 0101-1246 5065-9922 5063-6502 5067-6845 0101-1244 5067-6831
AG1535-4048EA 200.32 USD InStock
CAAG5067-1503 CAAG5068-0321 AG1535-4048 AG1400-3166 CAAG1535-4048 AG5063-6502 AG5068-0321 CAAG1535-4046 AG5067-4191 AG0101-0376 AG0101-1245 CAAG0101-1248 AG0101-1244 AG0101-1243 AG0101-1242 CAAG5067-4191 CAAG0101-1245 CAAG1400-3166 CAAG0101-1244 CAAG0101-1246 CAAG0101-1243 AG5067-6831 CAAG5067-6831 CAAG0101-1242 AG0101-1248 AG5067-1503 AG0101-1246 CAAG5065-9922 AG5065-9922 CA0101-0376 CAAG5067-6845 AG5067-7031 CAAG5067-7033 AG5067-7032 CAAG5063-6502 CAAG5067-7032 CAAG5067-7031 AG5067-7033 AG1535-4046 AG5067-6845
CrossLab Manual Injection Valves and Supplies for HPLC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Industry-leading manual injection valves and related high-quality supplies.
  • Both analytical and preparative injection valves are available to fit different sample volumes and flow rates
  • Various rotor seal materials are available, including Vespel, Tefzel, and PEEK, to accommodate a range of sample pH
  • Choice of stainless steel or PEEK flowpath
  • The right mix of factory-cut and finished sample loops of highest quality for your application needs
  • Color-coded manual syringes with a wide selection of volumes, improved scale readability, and optimal accuracy and precision
Industry-leading Agilent manual injection valves are designed to ensure trouble-free operation of your HPLC system. Valves feature patented 'Make-Before-Break' architecture that allows switching between LOAD and INJECT positions without interrupting flow, so more samples may be analyzed in less time. Injection valves are available with different position and port injections, and deliver effective operation under different pressures, to ensure you get the right injection valve for your system and application.
Injection valves are available with different position and port injections, and deliver effective operation under different pressures, to ensure you get the right injection valve for your system and application.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty