Would you like to log in again?
High Recovery Vials and Inserts, Agilent Technologies
Supplier: Agilent
5188-6592 5183-4510 5180-0841 5184-3554 5184-3554 5184-3556 5188-6591 9301-1388 5184-3551 5184-3551 5184-3550 5182-3454 5188-6591 5184-3556 5180-0844 5180-0844 5188-6591 5188-6594 5188-6572 5184-3552 5188-6572 9301-1388 5188-6593 5184-3550 5182-3454 5188-6592 5183-4510 5184-3555 5188-6593 5183-2030 5183-2030 5188-6594 5184-3552 5188-6592 5183-2073 5183-2073 5184-3555 5180-0841
AG5183-2073PK 226.95 USD InStock
AG5183-2073 CA76514-806 AG5183-2030 AG5182-3454 CA5184-3554 CA5188-6593 CA76514-920 CA5183-2073 CA5188-6594 CA5188-6572 CA5184-3556 AG9301-1388 CA5188-6591 CA5188-6592 CA5180-0844 CA76515-144 AG5183-4510 CA76514-916 CA76514-918 CA76514-814 101478-116 CAAG5180-0841 101478-118 CA76514-894 AG5184-3556 AG5184-3555 AG5184-3554 AG5188-6593 AG5180-0844 AG5184-3552 AG5188-6572 AG5188-6594 AG5180-0841 AG5184-3551 AG5184-3550 AG5188-6591 AG5188-6592 CA76508-168
High Recovery Vials and Inserts, Agilent Technologies
Vials
Extract the maximum possible amount of precious samples.
  • High recovery vials enable sample concentration and injection without transferring to microvolume inserts
  • Micro-sampling vials offer a lower cost alternative to high recovery vials
  • Conical inserts with polymer feet virtually eliminate dead volume and act as shock absorber for needle
  • Flat bottom inserts are fitted to vial dimensions
Vial inserts are the perfect fit for HPLC and GC instrument platforms across a wide variety of applications, including pharmaceutical, environmental, energy and fuels, forensics, materials science, biopharmaceutical, proteomics, and metabolomics.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty