Would you like to log in again?
Headspace Vials and Caps, Agilent Technologies
Supplier: Agilent
CrossLab
5190-2286 5182-0837 5190-2285 5067-0227 5188-6538 8010-0180 5182-0838 5185-5957 5190-9598 8010-0152 5183-4474 5190-2287 5188-6537 5190-2238 5190-2288 5182-0837 8010-0152 5190-6147 5188-2753 5190-2238 5182-0840 8010-0412 8010-0179 5182-0838 5182-0839 5185-5957 5190-2239 5183-4474 5067-0227 5067-0235 5188-2753 8010-0413 5190-2288 5188-6537 5182-0839 5191-5634 5183-4475 8010-0413 5190-2286 5190-9598 8010-0179 8010-0412 5188-6538 5188-5392 5191-5634 5067-0226 5188-5392 5190-2285 5183-4475 5067-0226 5182-0840 8010-0180 5190-2287 5190-2239
AG5182-0840PK 142.79 USD InStock
AG5182-0840 AG5185-5957 CAAG8010-0179 CA76514-864 CAAG5185-5957 CA76514-862 CA5188-6537 CA5188-6538 AG8010-0179 AG8010-0413 AG8010-0412 AG5182-0837 AG5182-0838 CA76508-190 CA8010-0180 AG5182-0839 CAAG5190-2238 CA5182-0840 AG5188-6537 CAAG5191-5634 AG5188-6538 AG5067-0227 AG5067-0226 AG5190-2285 CAAG8010-0152 CAAG5190-2287 AG5190-2287 AG5190-2286 AG5190-2288 AG8010-0152 CA8010-0413 AG5183-4475 CA8010-0412 AG5183-4474 CAAG5190-9598 CA76508-180 AG5188-5392 CA76508-134 CA76508-136 AG5190-9598 CA5190-2239 AG8010-0180 CA5067-0227 CA5067-0226 AG5190-2238 AG5190-2239 CA76514-794 103584-262 CA5190-2286 CA5190-2285 AG5188-2753 AG5191-5634 101401-416 CA5190-2288
Headspace Vials and Caps, Agilent Technologies
Vials
Engineered to prevent sample contamination, with caps that provide consistently secure seals.
  • Beveled top for maximum seal security
  • Available in flat bottom glass vial or rounded bottom glass vial designs
  • Optional pressure safety release cap at 45 psi
  • High-performance cap septa designed for higher temperature usage up to 300 °C
Crimp caps are most common however many customers use screw sample and application dependent. Headspace vials come in both flat bottom and round bottom styles for use in a wide variety of headspace instrument platforms. Headspace screw caps and crimp caps come in a variety of styles and materials including aluminum, bimetal, and steel. Headspace cap septa are available in PTFE/silicone, molded PTFE/butyl, and high-temperature options.
These headspace vials and caps are a perfect fit for a wide variety of applications, including pharmaceutical, environmental, energy and fuels, forensics, materials science, biopharmaceutical, proteomics, and metabolomics.
These headspace vials are constructed from type 33 and type 51 COE borosilicate glass, with low metal content, to protect your sample from destabilizing or leaching.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty