Would you like to log in again?
Peristaltic Pump Tubing for ICP-OES, Agilent Technologies
Supplier: Agilent
3710044400 3710043800 3710035200 3710068600 3710044400 3710044200 3710034500 3710068400 8003-0459 3710047000 3710069300 3710041700 3710043800 3710027200 3710044100 3710043700 3710057500 3710044200 3710034400 3710069000 3710044300 3710068400 3710069100 3710027300 8003-0459 3710068200 3710049000 3710034800 3710068500 3710046900 3710053700 3710049000 3710027200 3710041800 3710069100 3710034800 3710041800 3710053600 3710035000 3710068800 3710041700 3710043900 9910075900 9910075900 3710027300 3710043700 3710046900 3710035200 3710034600 5043-0063 3710068100 3710053700 3710044300 3710047000 3710041900 3710035000 5043-0063 3710068500 3710044000 3710044100 3710068600 3710068700 3710068000 3710069300 3710057500 3710034600 3710043500 3710068100 3710053600 3710043500 3710034400 3710034500 3710044000 3710069000 3710068300 3710068000 3710043900 3710068200 3710041900 3710068300 3710068800 3710068700
AG3710034800PK 100.38 USD InStock
AG3710034800 CA3710068800 CA3710035000 AG3710047000 CAAG3710069300 CAAG3710069100 CA5043-0063 AG3710043800 CAAG3710027300 AG3710069300 CAAG3710044000 CAAG3710044200 AG3710069100 CAAG3710044400 CA3710034500 AG3710034600 AG3710034400 AG3710053600 CAAG3710043800 CA3710046900 AG3710027300 CAAG3710068600 CAAG3710068400 CAAG3710068200 AG3710044100 AG3710041800 CAAG3710068000 AG3710044300 AG3710068700 CAAG3710053600 AG3710068500 AG3710068300 AG3710035000 CAAG3710047000 AG3710068100 AG8003-0459 AG3710035200 AG9910075900 CAAG3710041900 CAAG3710041700 AG3710057500 AG5043-0063 CA3710068100 AG3710049000 CA3710068300 CAAG3710057500 CAAG3710069000 AG3710043500 CA3710034800 AG3710043700 CA3710027200 AG3710043900 CAAG3710044100 AG3710069000 CAAG3710044300 AG3710053700 CA3710034400 CA3710034600 AG3710034500 CAAG3710043900 CAAG3710068700 AG3710027200 CAAG3710068500 AG3710044000 AG3710041900 AG3710044200 AG3710041700 AG3710044400 CAAG3710053700 AG3710068800 AG3710046900 AG3710068600 AG3710068400 CA3710049000 AG3710068200 CA3710035200 AG3710068000 CA9910075900 CAAG3710041800 CAAG3710043500 CAAG3710043700 CAAG8003-0459
Peristaltic Pump Tubing for ICP-OES, Agilent Technologies
Tubing Peristaltic Pump Tubing
PVC, Solvaflex, Marprene and FPM peristaltic pump tubing for ICP-OES.
  • Standard PVC suitable for all aqueous with medium concentrated acids or bases
  • PVC Solvaflex for common organic solvents such as kerosene, xylene, white spirit, alcohols, medium-highly concentrated acids and bases
  • Marprene tubing for ketone-based solvents such as MIBK and DIBK;FPM Fluoroelastomer Polymer tubing is suitable for gasoline, naphtha, toluene, xylene (aromatic hydrocarbons) and highly concentrated acids and bases
Agilent top-quality ICP-OES Peristaltic Pump Tubing allows you to find the appropriate product to handle any matrix. Agilent has tubing in the diameters and material required for your application: PVC, Solvaflex, FPM, or Marprene.
;Standard PVC works for all aqueous with medium concentrated acids or bases, PVC Solvaflex works for common organic solvents such as kerosene, xylene and white spirit. Marprene tubing is for ketone-based solvents such as MIBK and DIBK. FPM Fluoroelastomer Polymer tubing is for gasoline, naphtha, toluene, xylene (aromatic hydrocarbons) and highly concentrated acids and bases.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty