Would you like to log in again?
CrossLab Skimmer Bases for ICP-MS, Agilent Technologies
Supplier: Agilent
CrossLab
G1820-65435 G8400-60624 G3666-60301 G8400-60625 G3280-60381 G3280-60608 G3666-60601 G3280-60381 G3280-60621 G8400-60625 G3666-60401 G3280-60608 G1833-65590 G3666-60301 G3666-60311 G1833-65591 G8400-60628 G8400-60626 G1820-65435 G3666-60311 G8400-60628 G8400-60627 G8400-60624 G1833-65591 G8400-60627 G8400-60634 G8400-60634 G3666-60601 G3666-60401 G1833-65590 G3280-60621 G8400-60626
AGG3666-60301EA 651.82 USD InStock
AGG3666-60301 CAAGG328060608 AGG3666-60401 AGG3666-60601 AGG8400-60634 CAAGG328060621 CAAGG366660601 CAAGG366660301 CAAGG182065435 CAAGG366660401 AGG1820-65435 CAAGG840060624 AGG3280-60608 AGG1833-65591 AGG3280-60621 AGG1833-65590 AGG3280-60381 AGG3666-60311 AGG8400-60625 AGG8400-60624 CAAGG183365590 CAAGG183365591 CAAGG366660311 CAAGG840060634 CAAGG840060625 CAAGG328060381 CAAGG840060626 CAAGG840060627 CAAGG840060628 AGG8400-60628 AGG8400-60627 AGG8400-60626
CrossLab Skimmer Bases for ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Agilent stainless steel and brass skimmer bases optimize the temperature of the skimmer cone tip.
  • Agilent 7700, 7800, 7900, 8800, 8900 ICP-MS skimmer cone base are avilable in either stainless steel or brass
  • The operating temperature of the skimmer cone tip is controlled by the skimmer base material
  • The stainless steel base is used for nickel (Ni) skimmer cones. The platinum skimmer cone should only be used with the brass skimmer base
  • The ion extraction lenses (x-lens or s-lens) are fixed to the skimmer base
  • The correct type for lens configuration and ICP-MS model (7700, 7800, 7900, 8800, 8900) is required
The ion extraction lenses are fixed to the skimmer base;the correct type for lens configuration and ICP-MS model is required.
The skimmer base ensures the correct operating temperature of the skimmer cone to prevent damage by overheating or increased drift due to excess matrix deposition at the tip if the cone runs too cool. Firmly install the sampling and the skimmer cones on the skimmer base. If the interface cones are not tightly installed on the base, there may be insufficient cooling, which can degrade analytical performance
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty