Would you like to log in again?
Crosslab Packed GC Columns, Agilent Technologies
Supplier: Agilent
J&W
CP81072 G3591-81135 G3591-81156 G3591-81209 G3591-81012 G3591-81002 CP2059 G3591-81022 G3591-81023 G3591-81158 G3591-81096 G3591-81037 G3591-80020 G3591-81121 G3591-80013 G3591-81142 G3591-81020 G3591-81017 G3591-80036 G3591-81212 G3591-81045 G3591-81026 G3591-81210 G3591-80003 G3591-70001 G3591-80017 G3591-70013 G3591-80006 CP2059 G3591-80043 G3591-81121 G3591-80157 G3591-70001 G3591-81013 G3591-70013 G3591-80033 G3591-81082 CP2065 G3591-80017 G3591-80033 G3591-81003 CP81071 G3591-81162 G3591-81011 G3591-81097 G3591-82000 G3591-80021 G3591-81158 G3591-80158 G3591-81003 G3591-81085 G3591-80158 G3591-81011 G3591-81037 G3591-81064 G3591-81023 G3591-81212 G3591-80003 G3591-81141 G3591-81012 G3591-81142 G3591-81187 G3591-82000 G3591-81161 G3591-81004 G3591-81156 G3591-80006 G3591-80013 G3591-81135 G3591-81214 G3591-81085 G3591-81210 G3591-81013 G3591-81082 G3591-81209 G3591-81026 G3591-82001 G3591-80043 CP81071 G3591-81017 G3591-82020 G3591-81002 G3591-81187 G3591-81181 G3591-81020 G3591-81141 CP2065 G3591-80021 G3591-82020 G3591-81160 G3591-81181 G3591-81000 G3591-82001 G3591-81006 CP81072 G3591-81097 G3591-81004 G3591-81162 G3591-81214 G3591-81022 G3591-80157 G3591-81161 G3591-81006 G3591-81000 G3591-81064 G3591-80020 G3591-81096 G3591-81045 G3591-81160 G3591-80036
AGG3591-81026EA 752.49 USD InStock
CAAGG359181006 AGG3591-81026 AGG3591-81023 AGG3591-81142 AGG3591-81022 AGG3591-81020 AGG3591-81141 CAAGG359181096 CAAGG359181013 AGG3591-70001 CAAGG359181135 CAAGG359181097 CAAGCP2065 CAAGG359180043 CAAGG359181011 CAAGG359181012 CAAGCP2059 CAAGG359181017 AGG3591-81017 AGG3591-81135 AGG3591-81012 AGG3591-80043 AGG3591-81013 AGG3591-81011 AGG3591-81096 AGG3591-81097 AGCP2059 AGG3591-70013 CAAGG359181026 CAAGG359181020 CAAGG359181141 CAAGG359181142 CAAGG359181022 CAAGG359181023 AGG3591-80157 AGG3591-80036 AGG3591-81006 AGG3591-81003 AGG3591-80158 AGG3591-81004 AGG3591-81002 AGG3591-81121 AGG3591-80033 AGG3591-81000 AGG3591-81085 CAAGG359181156 CAAGG359181158 CAAGG359181037 CAAGG359182000 CAAGG359182001 AGG3591-80021 AGG3591-80020 AGG3591-81082 CAAGG359181160 CAAGG359181161 CAAGG359181162 CAAGG359181045 AGG3591-80017 CAAGG359181209 AGG3591-80013 AGG3591-81187 AGG3591-81064 CAAGG359182020 CAAGG359181212 CAAGG359180003 CAAGG359181214 CAAGG359181210 AGG3591-80006 AGG3591-80003 AGG3591-81214 CAAGG359180006 AGG3591-81212 AGG3591-81210 AGG3591-82020 CAAGG359181181 AGG3591-81181 CAAGG359180013 CAAGG359181064 CAAGG359181187 CAAGG359180017 AGG3591-81045 CAAGG359170013 AGCP2065 AGG3591-81162 AGG3591-81160 AGG3591-81161 CAAGG359180020 AGG3591-81209 CAAGG359180021 CAAGCP81072 CAAGCP81071 AGG3591-81158 AGG3591-81037 AGG3591-81156 AGG3591-82000 AGG3591-82001 CAAGG359170001 CAAGG359181082 CAAGG359181085 CAAGG359181002 CAAGG359181003 CAAGG359180036 CAAGG359180157 AGCP81071 CAAGG359181004 CAAGG359180158 AGCP81072 CAAGG359181121 CAAGG359181000 CAAGG359180033
Crosslab Packed GC Columns, Agilent Technologies
Chromatography Columns
Excellent column-to-column reproducibility, available in a variety of tubing materials, support materials, and liquid phases.
  • UltiMetal deactivated stainless steel tubing promotes sharp peaks with less peak tailing for improved data accuracy
  • High-efficiency packing ensures excellent reproducibility in pressure drop, flow characteristics, and separation performance resulting in less system readjustment after column installation
  • Select from a range of tubing materials, including stainless steel, UltiMetal deactivated stainless steel, nickel, glass, copper, and PTFE – plus hundreds of stationary phases, packings, and supports;Create your own custom configurations
  • Column configurations are available for all major GC brands, past and present, for worry-free compatibility
Agilent J&W Packed GC Columns are designed and manufactured to offer excellent and reproducible performance for all sample types associated with packed column separations, most important in the hydrocarbon processing industry. The highly efficient and rigorous packing technology used in Agilent J&W packed GC columns assures column-to-column reproducibility and efficiency, while the UltiMetal deactivated stainless steel tubing allows for improved inertness and peak shape performance.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty