Would you like to log in again?
CrossLab Demountable Torch Supplies for 5100/5110 and 5800/5900 ICP-OES, Agilent Technologies
Supplier: Agilent
CrossLab
G8010-67000 G8014-60023 G8010-60227 G8010-68021 G8010-60227 G8010-67000 G8010-68021 G8014-60023
AGG8010-60227EA 97.63 USD InStock
CAAGG801067000 CAAGG801060227 AGG8010-60227 AGG8010-67000 AGG8014-60023 AGG8010-68021 CAAGG801068021 CAAGG801460023
CrossLab Demountable Torch Supplies for 5100/5110 and 5800/5900 ICP-OES, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Injectors, outer tube sets, and other supplies for the fully demountable torch.
  • Fully demountable torch provides the flexibility of having a variety of injector sizes and materials to handle difficult and diverse application demands
  • Ease of maintenence with a tool free removable injector, all with no optimization required
  • New organics outer tube set is made of high purity quartz providing longer lifetime when using organic solvents
  • The vertical torch orientation provides the ultimate configuration for determining tough samples
  • Handles even the toughest samples and achieves unmatched long term stability
  • The plug-and-play torch eliminates the torch alignment process
  • Ensures fast start-up and reproducible results with each operator
The new fully demountable torch for the 5000 Series ICP-OES can be used with a variety of injector sizes and materials to analyze any sample matrix. Outer tube sets are available in axial and radial configurations with an organics option available. The organics outer tube set extends the service life of the tube set when used with organic applications. The fully demountable torch features a removable injector to simplify torch maintenance and allows quick changeover for diverse sample matrices. The complexity of changing the injector when switching applications or cleaning has never been easier.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty