CB3754-112
CB3754-032
CB3754-001
CB3754-130
CB3754-157
CB3754-014
BDH0611-4LCS
4240.27
USD
InStock
BDH0611-4L
BDH0611-200L
BDH0611-450ML
BDH0611-20L
BDH0611-2.5L
BDH0611-56L
Cap B 50:20:30 Anhydrous ACN/Acetic Anhydride/Lutidine for biosynthesis, VWR Chemicals BDH®
Capping Reagent
BDH Biosynthesis Grade Reagents are designed for Biosynthesis research and production. Sizing available to fit your needs, from benchtop synthesizers to bulk production.
- Custom Blends and Bulk Sizes available upon request.