Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethyl sulfoxide ≥99.9% ACS, VWR Chemicals BDH®

Supplier: VWR International
Synonyms: Methyl sulfoxideDMSODimethyl(oxido)sulfurMethyl sulphoxideMethanesulphinylmethaneDimethyl(oxido)sulphurMethanesulfinylmethane

BDHVBDH1115-4LP BDHVBDH1115-1LP
BDH1115-4LPCS 1790.31 USD InStock
BDH1115-4LP BDH1115-1LP
Dimethyl sulfoxide ≥99.9% ACS, VWR Chemicals BDH®
Dimethyl sulphoxide


Formula: (CH₃)₂SO
MW: 78.14 g/mol
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: 18.5 °C
Density: 1.101 g/cm³ (20 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://us-legacy.vwr.com/stibo/web/std.lang.all/77/31/2297731.jpg
Order Now
Specification Test Results
Appearance Clear, colorless liquid
Water (K.F.) Less than 0.10%
Residue Less than 0.01% (0.0010g/91mL)
GC Purity Greater than 99.9%
Color (APHA) Less than 10
Titrable acid Less than 1µeq/g (Not more than 5.0mL of of 0.01N sodium hydroxide)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty