Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Propanol ≥70%, VWR Chemicals BDH®

Supplier: VWR International
Danger
Synonyms: Isopropyl alcoholIsopropyl Rubbing Alcoholsec-PropanolIsopropyl rubbing alcohol2-Hydroxy propanesec-Propyl AlcoholDimethyl carbinol2-HydroxypropanePropane, 2-hydroxy-Propan-2-olIPAIsopropanol

BDHVBDH1131-204L BDHVBDH1131-5GL BDHVBDH1131-1LP BDHVBDH1131-4LP
BDH1131-5GLEA 344.05 USD InStock
BDH1131-5GL BDH1131-204L BDH1131-1LP BDH1131-4LP
2-Propanol ≥70%, VWR Chemicals BDH®
2-Propanol
Clear, colorless liquid.
Meets ACS specifications for general use.


Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Density: 0.8…0.92 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1987
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/19/69/731969.jpg
Order Now
Specification Test Results
Appearance Clear, colorless liquid
Isopropyl Alcohol 68.0-72.0%
Water 28.0-32.0%
Specific gravity @ 20ºC 0.863-0.883
Color (APHA) Less than 10

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty