Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Propanol ≥99.5% ACS, VWR Chemicals BDH®

Supplier: VWR International
Danger
Synonyms: Isopropyl alcoholIsopropyl Rubbing Alcoholsec-PropanolIsopropyl rubbing alcohol2-Hydroxy propanesec-Propyl AlcoholDimethyl carbinol2-HydroxypropanePropane, 2-hydroxy-Propan-2-olIPAIsopropanol

BDHVBDH1133-4LG BDHVBDH1133-19L BDHVBDH1133-1LP BDHVBDH1133-5GL BDHVBDH1133-204L BDHVBDH1133-4LP
BDH1133-19LEA 287.66 USD InStock
BDH1133-19L BDH1133-204L BDH1133-1LP BDH1133-4LG BDH1133-4LP BDH1133-5GL
2-Propanol ≥99.5% ACS, VWR Chemicals BDH®
2-Propanol
Suitable for histology and cytology applications. Soluble in water. Meets ACS Specifications for general use.


Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82 °C (1013 hPa)
Melting Pt: –89 °C
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://us-legacy.vwr.com/stibo/web/std.lang.all/19/69/731969.jpg
Order Now
Specification Test Results
Appearance Clear
Water (K.F.) Less than 0.20%
Residue Less than 0.001% (0.0010 g/128 mL)
GC Purity Greater than 99.5%
Solubility in water Passes test (Solution should be as clear as an equal volume of water.)
Color (APHA) Less than 10
Titrable acid or base Less than 0.1 meq/g (No more than 0.10 mL of either 0.01N sodium hydroxide or 0.01N hydochloric acid)
Carbonyl compounds Less than 0.002% each (as acetone or propionaldehyde)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty