Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trifluoroacetic acid ≥99.8%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®

Supplier: VWR International
Danger
Synonyms: TFA

BDH15311.100 BDH15311.100
BDH15311.100CS 2374.94 USD InStock
BDH15311.100 CABDH15311.100
Trifluoroacetic acid ≥99.8%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Trifluoroacetic acid


Formula: CF₃COOH
MW: 114.02 g/mol
Boiling Pt: 71.8 °C (1013 hPa)
Melting Pt: –15 °C
Density: 1.490 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
EINECS: 200-929-3
UN: 2699
ADR: 8,I
Merck Index: 12,09812
https://us-legacy.vwr.com/stibo/web/std.lang.all/83/92/728392.jpg
Order Now
Specification Test Results
Assay by Titration Min. 99.8 %
Color Max. 10 APHA
Transmittance (210 nm) 0.1% aq. soln Min. 30 %
Transmittance (220 nm) 0.1% aq. soln Min. 60 %
Transmittance (230 nm) 0.1% aq. soln Min. 85 %
Transmittance (240 nm) 0.1% aq. soln Min. 95 %
Transmittance (250 nm) 0.1% aq. soln Min. 99 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty