Synonyms:
Lunar causticNitric acid silver saltSilver mononitrate
BDHVBDH7214-5
BDHVBDH7214-1
BDHVBDH7214-4
BDHVBDH7214-7
BDHVBDH7214-2
BDH7214-7EA
1736.86
USD
InStock
BDH7214-7
BDH7214-5
BDH7214-4
BDH7214-2
BDH7214-1
Silver nitrate 0.1 N in aqueous solution, VWR Chemicals BDH®
Silver nitrate
Suitable as a titrant. Made with deionized water.