Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium chloride 99.0-100.5% ACS, VWR Chemicals BDH®

Supplier: VWR International
Synonyms: Potassiumchloride

BDHVBDH9258-500G BDHVBDH9258-2.5KG
BDH9258-500GCS 868.38 USD InStock
BDH9258-500G BDH9258-2.5KG
Potassium chloride 99.0-100.5% ACS, VWR Chemicals BDH®
Potassium chloride
Meets reagent specifications for testing USP/NF monographs.

Colorless or white solid


Formula: KCl
MW: 74.55 g/mol
Boiling Pt: 1420 °C (1013 hPa)
Melting Pt: 773 °C
Density: 1.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Merck Index: 13,07704
Order Now
Specification Test Results
Barium Test PASS
Bromide <= 0.01 %
Calcium <= 0.002 %
Chlorate & Nitrate (as NO3) <= 0.003 %
Heavy Metals <= 0.0005 %
Insolubles <= 0.005 %
Iodide <= 0.002 %
Iron <= 0.0003 %
Magnesium <= 0.001 %
pH (5%, Water) @25C 5.4 - 8.6
Phosphate <= 0.0005 %
Purity 99.0 - 100.5 %
Sodium <= 0.005 %
Sulfate <= 0.001 %
Meets reagent specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty