Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Triethylamine ≥99.5%, BioSyn™ for HPLC, Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: TEA(Diethylamino)ethane

17924-1L 17924-1L 17924-10X2ML 17924-10X2ML
BJ17924-10X2MLCS 255 USD InStock
BJ17924-10X2ML CABJ17924102ML CABJ17924-1L BJ17924-1L
Triethylamine ≥99.5%, BioSyn™ for HPLC, Fluka™
Triethylamine
Beilstein Number: 605283
RTECS Number: YE0175000
UNSPSC Code: 12352116


Formula: (C₂H₅)₃N
MW: 101.19 g/mol
Boiling Pt: 90 °C
Melting Pt: -115 °C
Density: 0.73 g/cm³ (20 °C)
Flash Pt: -11 °C
Storage Temperature: Ambient
MDL Number: MFCD00009051
CAS Number: 121-44-8
EINECS: 204-469-4
UN: 1296
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/92/82/729282.jpg
Order Now
Specification Test Results
Assay (GC) min. 99.5 %
Suitability test (HPLC) complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty